Cell Detachment Solutions, DetachKit 2, PromoCell
Supplier: Promocell
C-41202 C-41212 C-41222
10175-294EA 99.7 CAD
10175-294 10175-298 10175-302
Cell Detachment Solutions, DetachKit 2, PromoCell
Cell Culture Supplements and Additives
This DetachKit 2 is used for very gentle, safe and efficient detachment of primary human cells in routine subculturing.
  • Can be used for primary animal cells and cell lines
  • Valuable for more sensitive cell types
  • Comes with a Trypsin/EDTA ratio of 0.025%/0.01%
Each DetachKit consists of three components: HEPES BSS (HEPES buffered Balanced Salt Solution), Trypsin/EDTA Solution and TNS (Trypsin Neutralization Solution).
HEPES BSS contains 30 mM HEPES, D-Glucose, NaCl, KCl, Na-Phosphate and Phenol Red. TNS contains 0.05% Trypsin Inhibitor from soybean and 0.1% Bovine Serum Albumin.