Trypsin EDTA Solution, PromoCell
Supplier: Promocell
C-41000
C-41002
C-41010
C-41012
C-41020
C-41022
10172-544EA
54.46
CAD
10172-544
10172-546
10172-548
10172-550
10172-552
10172-554
Trypsin EDTA Solution, PromoCell
Cell Culture Supplements and Additives
Trypsin is the most widely employed enzyme used for cell detachment.
- Available in different sizes
- Store at –20 °C in the dark
- Thawed solutions can be stored at 4 to 8 °C
PromoCell's Trypsin Solutions exhibit standardized enzyme activity and contain trypsin from extra-pure lots.
All lots have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).
Caution: The products are for in vitro research use only and not for diagnostic or therapeutic procedures.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...