Light Emitting Diodes (LED), Green, Eisco Scientific

Supplier: Eisco Scientific
PH1019B
470230-384PK 10.7 CAD
470230-384
Light Emitting Diodes (LED), Green, Eisco Scientific
Lights LED Lights, Lamps and Bulbs
Standard green LED indicattor with diffused lens

  • Length,1.5"
  • Diameter, 0.196"
  • Metal connection prongs
  • Maximum 3 V, DC

These green light emitting plastic diodes (LEDs) are ideal for use in either PCB mount or panel mount calibrations on a circuit board. The LEDs provide a green light source while using very little energy.

Packaging: Pack of 10.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR