Round Flash Lamp Bulbs, 3.5 V, Eisco Scientific

Supplier: Eisco Scientific
PH1014B
470230-378PK 10.75 CAD
470230-378
Round Flash Lamp Bulbs, 3.5 V, Eisco Scientific
Lights Incandescent Lights, Lamps and Bulbs
Suitable for flash lamps, spotlights and panel lamps. 

  • Size E10 screw base
  • For use with basic circuit kits
  • Works with flash lamps, spot lamps and panel lamps
  • 3.5 V with M.E.S. cap

Packaging: Supplied in pack of 10.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR