Ward's® Live Cave Roaches
Supplier: Avantor
470315-540EA 41 CAD
470315-540 470315-542 470315-544
Ward's® Live Cave Roaches
Specimens Live Specimens Animals - Living Material
Cave roaches are widely used in labs for physiological, behavioral, structural, and developmental studies.
  • Blaberus discoidalis
Specimens come from cultures bred in the lab environment.
Ordering information: These products contain living or perishable materials.
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Packaging: Specimens are packed in either a 16 Oz. or 32 Oz. jar depending on sizing.