IncuShaker Shaking Incubator Series
Supplier: Ward's
H1010-P-MP H1010-P-SP H1010-SH H2300-REG
470319-024EA 1598.65 CAD
470319-024 470319-026 470319-028 470319-038
IncuShaker Shaking Incubator Series
Shakers Shaking Incubators
Digitally regulated temperature with precise mixing control.
  • Great for cell culture or bacterial cell growth
  • Various sizes and configurations available
  • Two years warranty
The Incu-Shaker™ series of shaking incubators provide digitally regulated temperature with precisely controlled, circular (orbital) mixing. The IncuShaker™ Mini is the most compact shaking incubator in its class. Despite the remarkably small footprint, a large work space accepts several platforms and a variety of flasks, tubes and other common vessels. It is available in both anMore Product Information