Corepoint Scientific™ Refrigerators, Horizon Scientific
Supplier: Horizon Scientific
PR101WWWG/0 PR101WWW/0
76335-186EA 15604.71 CAD
76335-186 76335-188
Corepoint Scientific™ Refrigerators, Horizon Scientific
Refrigerators
These compact laboratory refrigerators offer professional cold storage for scientific and clinical settings.
  • Microprocessor temperature control
  • Integrated temperature alarms
  • Keyed door locks
  • Probe access
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
Units come equipped with forced draft circulation and precise temperature monitoring through microprocessor control. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Certifications: *Select models ENERGY STAR certified: 76335-186 and 76335-188
Ordering information: All units come with a three year parts and labor warranty, plus an additional two year compressor parts warranty.