VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
76307-462 76307-464 76307-466 76307-468 76307-461
76307-462BX 73.85 CAD
76307-462 76307-464 76307-466 76307-468 76307-461
VWR® Nitrile Examination Gloves, Black
Gloves
Advanced nitrile material formulation features added thickness to provide excellent barrier protection. Gloves are produced in ISO certified facilities.
  • Powder-free and latex-free
  • Beaded cuffs
  • Ambidextrous
  • Thickness: 5.3 mil (Finger), 4.5 mil (Palm), 3.5 mil (Cuff)
The formulation with a finger textured surface for excellent barrier protection, durability, and abrasion resistance during extended wear. The black color offers a sleek look and versatility for various applications.
Certifications: FDA 510(k) Approved as a Medical Device. FentanylMore Product Information