Ultrasonic Homogenizers Sonic Ruptor 400, Omni International
Supplier: Omni International
18-000-115
CA11012-236EA 18885.19 CAD
CA11012-236
Ultrasonic Homogenizers Sonic Ruptor 400, Omni International
Homogenizers Ultrasonic Cell Disruptors
Sonic Ruptor 400 Ultrasonic Homogenizer offers precision engineering with all the features necessary to create a total system for ultrasonic disruption. It can disintegrate most cells, bacteria, spore or tissue.
  • Variable power up to 400 W
  • Automatic tuning for easy interchange of probe tips
  • Pulse mode power allows samples to be processed while limiting temperature rise
  • Integrated timer and adjustable power
  • Wide range of accessories including sound abatement chambers and continuous flow chambers
It can also prepare an emulsion down to 1/100 of a micron, homogenize immiscible liquids, accelerate enzymatic and chemical reactions, stimulate bacterial activity, disperse solids in liquids and degas liquids.
It enables full amplitude control from 0 to 100% and is compatible with sonicator horns ranging from 1/8 to 1".
Ordering information: Processing tips, stand assembly and accessories sold separately. Unit supplied with 400 W control unit, transducer, tool kit and instruction manual.