Amino Labs Engineer-It Kit™
Supplier: AMINO LABS NORTH INCORPORATED CA SE
WWE004-F_CA WWE004-G_CA WWE004-S_CA WWE004-SR_CA
CA470318-838EA 666.09 CAD
CA470318-838 CA470318-840 CA470318-842 CA470318-844 CA470318-846
Amino Labs Engineer-It Kit™
Educational Materials Biology Educational Materials AP Biology Learning Activities
Full STEAM ahead with the Engineer-It Kit™; grow and engineer bacteria to create color pigments.
  • Safety First! The kit uses a non-pathogenic strain of E. coli (K-12)
  • No need to purchase any additional reagents
  • Recommended for use with Amino Labs DNA Playground™ (available separately)
  • Kit complements several Next Generation Science Standards (NGSS)
Learn genetic engineering hands-on using the Engineer-It Kit™. The end result is that you have grown and engineered bacteria to produce something for you - a colorful bio-pigment. 1-color Engineer-ItMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable