Amino Labs Extract-It Kit™
Supplier: AMINO LABS NORTH INCORPORATED CA SE
WWG005-G_CA WWG005-S_CA
CA470318-728EA 487.49 CAD
CA470318-728 CA470318-730
Amino Labs Extract-It Kit™
Educational Materials Biology Educational Materials AP Biology Learning Activities
Extract-It Kit™ lets you extract the protein made by the engineered bacteria you provide so you can use it outside of the system.
  • Kit utilizes user-supplied engineered cells for the extraction
  • Great next step after completing the Engineer-It™ or RGB Kit™ (also available)
  • Recommended for use with Amino Labs DNA Playground™ (available separately)
The Extract-It Kit™ allows you to collect the protein created by your DNA program plasmid (for example, a colored protein) so that you can use it elsewhere.
In this kit students will: Culture bacteria toMore Product Information