Wringers, Contec®
Supplier: Contec
2756 2738 2690 2689
33502-260EA 2188.82 CAD
33502-260 89198-850 89198-854 76199-888 76199-932
Wringers, Contec®
Mops
A variety of wringers designed for use with curved and flat mops.
  • Stainless steel and polypropylene wringers for a variety of Contec® bucket systems
  • Can be used with curved, flat, and string mops
  • Fully autoclavable
These wringers fit on Contec® 3.9 gallon (15 l), 5 gallon (20 l) , and 6.5 gallon (25 l) bucket systems. All wringers are compatible with a wide variety of disinfectants.