Amino Labs Heat-It Kit™
Supplier: AMINO LABS NORTH INCORPORATED CA SE
WWE009-S_CA WWE009-G_CA
CA470318-712EA 196.1 CAD
CA470318-712 CA470318-860
Amino Labs Heat-It Kit™
Educational Materials Biology Educational Materials AP Biology Learning Activities
Engineer cells with a DNA program where gene expression can be manually controlled with temperature.
  • Safety First! The kit uses a non-pathogenic strain of E. coli (K-12)
  • Recommended for use with Amino Labs DNA Playground™ (available separately)
The Heat-It Kit™ lets you grow and engineer bacteria with a DNA plasmid that can be controlled with heat. Decide when you want the DNA program to turn on by choosing the temperature you grow the bacteria at. Turn the DNA program on and off to create tree ring patterns in the growth.
In this kit, studentsMore Product Information