Polywipe® Light Wipes, Contec®
Supplier: Contec
Polywipe®
7070-99
68300-554CS 778.74 CAD
68300-554
Polywipe® Light Wipes, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Polywipe Light Wipes are unlaundered standard weight interlock knit polyester wipes with knife-cut edges that can be used in less critical applications.
  • Recommended for ISO Class 6 to 8 environments
  • Knitted 100% polyester fabric for low levels of particles and fibers
  • Soft edge is ideal for use on surfaces which are vulnerable to scratching
  • Cost effective for equipment wipe down and general cleaning
  • Durable no run interlock knit construction keeps the fabric from unraveling
  • Abrasion and chemical resistant
The durable, no-run interlock knit construction makes it ideal for equipment wipe down and general cleaning. It is available in different sizes and put-ups.