Amino Labs Junior R&D Kits™
Supplier: AMINO LABS NORTH INCORPORATED CA SE
WWE014-AMP_CA WWE014-CHLORCA
CA470318-714EA 320.99 CAD
CA470318-714 CA470318-716 CA470318-718 CA470318-720 CA470318-722
Amino Labs Junior R&D Kits™
Educational Materials Biology Educational Materials Genetics Learning Activities
This kit enables a junior researcher to complete DNA transformations with their own engineered DNA and cells.
  • Kit utilizes user-supplied engineered cells for the extraction
  • Great next step after completing the Engineer-It™ or RGB Kit™ (also available)
  • Recommended for use with Amino Labs DNA Playground™ (available separately)
Have you purchased or acquired a DNA plasmid that you'd like to put into E. coli bacteria? Have you completed some ligations and want to transform E. coli with your ligated products? Then these kits are for you! You supply the cells and DNA plasmid, and simply select the kit with the type of antibiotic you need for your experiments, and get ready for transitioning to your own R&D project.
Additional equipment required but not included: Microwave or hot-plate, and Amino Labs DNA Playground™ (In the absence of a DNA Playground™, you will also need an ice bath, water bath, thermometer, timer, and Incubator).
Ordering information: Due to popular demand, we recommend you place your order at least a week in advance.
Delivery information: Though kits will ship at ambient temperature, some components will require refrigeration upon arrival. Refer to instructions.