Nunc™ Cell Culture Flasks, Thermo Scientific
Supplier: Thermo Fisher Scientific
132703 132704 132705 132706 132707 132708 159920 153732 156502 178883 178905 156340 156367 156499 169900 156800 159926 178983 159933 159934 159910 156472 132703 132704 132705 132706 132707 132708 132097 132098 178885 178985
89020-032CS 1492.95 CAD
89020-032 89020-034 89020-036 89020-038 89020-040 89020-042 CA12777-958 CA15708-094 CA15708-096 CA15708-097 CA15708-101 CA15708-103 CA15708-106 CA15708-120 CA15708-130 CA15708-134 CA73521-146 CA73521-148 CA73521-150 CA73521-200 CA73521-358 CA73521-360 CA12777-960 CA15708-124 CA89020-032 CA89020-034 CA89020-036 CA89020-038 CA89020-040 CA89020-042 CA30617-428 CA30617-430 490012-436 10000-980 10000-982
Nunc™ Cell Culture Flasks, Thermo Scientific
Flasks Cell Culture Flasks
These Nunc™ cell culture flasks are designed for culture consistency, cell health, and reproducibility. Choose from a variety of surfaces and sizes with culture areas ranging from 25 to 500 cm² to suit your specific applications and cell types.
  • Nunclon Delta certification including testing on four different cell types is provided in each case
  • Barcoded 175 cm² flasks (*) with large Code 128 are compatible with automation
  • Sterilized by gamma irradiation and certified to be non-pyrogenic
Flasks treated with the proprietary Nunclon™ Delta surfaceMore Product Information