Mini Magnetic Stirrers
Supplier: Heathrow Scientific
120155 120595
470316-808CS 2186.7 CAD
470316-808 470316-818
Mini Magnetic Stirrers
Stirrers Magnetic Stirrers
Economical solution made for ultimate function with optimal magnetic coupling to ensure dependable stirring efficiency.
  • Made from an ABS and PC blend that is chemical resistant making the unit easy to clean
  • Low-profile design is lightweight for transporting with ease and the small footprint saves valuable space on a bench top
  • Quiet operation with low vibration which helps users stay focused by reducing noise in the lab
  • Two high quality stir bars (35×9 mm) are included to ensure maximum stirring performance
A powerful magnetic coupling of the high-quality stir bar and the unit ensures consistent accurate alignment which significantly reduces spin-outs during acceleration or deceleration of spin speeds and ensure maximum stirring performance, every time.
Ideal for routine stirring applications in general research, life science, industrial, quality analysis, and clinical lab settings.
Certifications: Intertek, CE, UKCA, CB Scheme, RoHS 2, and WEEE compliant
Delivery information: Includes 1 Mini magnetic stirrer, 2 stir bars, 1 Low voltage, double insulated power adapter with 4 interchangeable plugs. Supplied with 5 years warranty.