Ward's® MagFuge® Multi-Function High-Speed Centrifuge Bundle
Supplier: Heathrow Scientific
120581_SW 120582_SW
470316-830CS 4036.9 CAD
470316-830 470316-832
Ward's® MagFuge® Multi-Function High-Speed Centrifuge Bundle
Centrifuges Microcentrifuges
The MagFuge® is the industry’s first high-speed centrifuge and magnetic stirrer combined into one unit.
  • Five year warranty
  • Low-profile design is compact and has a small footprint
  • Easy to use back-lit digital panel and digital control
  • Rotor sensing technology
  • Low voltage insulated power adapter with four cord sets
The innovative design enables a substantial cost savings for laboratories by not having to purchase two individual pieces of equipment because the MagFuge® does it all.
The high-speed centrifuge is designed to give maximum speed and flexibility and is comfortable and easy to use. Safety features ensure it will not operate when the lid is open, a rotor is jammed, or the unit is out of balance.
Comes with rotors that can accommodate 12×1.5/2.0 ml tubes or 6×5 ml tubes. The magnetic stirrer offers a powerful magnetic coupling that ensures consistent, accurate alignment of the stirring bar that significantly reduces the chance of spin-outs. The stirrer can accommodate up to three liters of liquid.
Certifications: Meets all global certifications: CE, SGS, RoHS2, WEEE.
Accessories information: Quick setup of the MagFuge® centrifuge/stirrer with tool free rotor exhange. The two tube rotors include adapters to accommodate 1.5, 2.0, and 5 ml tubes. High-quality rare-earth magnets are used in the stirrer rotor to ensure the strongest magnetic coupling for a consistent and accurate alignment of the stir bar, which significantly reduces the chance of spin-outs. The silicone mat helps prevent spills and keeps vessel stable.
Delivery information: Supplied with MagFuge™, 2 stirring bars; 2 centrifuge rotors (1.5/2.0 ml, 5 ml); 1 stirrer rotor; 2 silicone lid mats (for use with stirrer); 1 low voltage, insulated power adaptor with four cord sets; 12×0.2 ml tube adaptors, 12×0.5 ml tube adaptors.