Roto-Therm™ Incubating Rotating Mixer, Benchmark Scientific
Supplier: BENCHMARK SCIENTIFIC CA
H2020 H2024
CA76333-866EA 13433.49 CAD
CA76333-866 CA76333-868
Roto-Therm™ Incubating Rotating Mixer, Benchmark Scientific
Shakers Rotating/Rocking Shakers
Roto-Therm™ series of incubated rotating mixers provide a wide range of mixing capabilities. They are suitable for carrying out an array of laboratory applications including resuspension, protein binding, nucleic acid purification, histochemistry, hybridization.
  • Rotated mixing with temperature control
  • Includes tube holders for common tube sizes
  • Available with variable speed control and advanced shaking/mixing modes: mix (oscillating), pause, rock
The mixers include interchangeable tube holders for accomomodating all common tube sizes from 0.5 to 50 ml.More Product Information