IKA PETTE fix and vario, Single Channel Pipettes, IKA WORKS
Supplier: IKA Works
0020011220 0020011221 0020011222 0020011223 0020011224 0020011225 0020011226 0020011227 0020011228 0020011210 0020011211 0020011213 0020011214 0020011215 0020011216 0020011217 0020011218 0020011219
76329-760EA 403.77 CAD
76329-760 76329-762 76329-764 76329-766 76329-768 76329-770 76329-772 76329-774 76329-776 76329-794 76329-796 76329-798 76329-800 76329-802 76329-804 76329-806 76329-808 76329-810
IKA PETTE fix and vario, Single Channel Pipettes, IKA WORKS
Pipettes Micropipettes
An ergonomic feel combined with uncompromising precise performance: the IKA single fix and vario pipettes can be used for many applications in the modern laboratory.
  • Exchangeable grips in different shapes and materials
  • Tip cone and piston are coated with a high quality diamind-like carbon (DLC)
  • Large display which shows set volumes in easily read characters
  • User-friendly IKA software
  • Simple maintenance and adjustment
Color coding simplifies the rapid selection of a suitable pipette. Resistance to impact, UV and chemicals ensures that the tip cone provides long-term reliable dosing. The PETTE fix and vario are fully autoclavable, therefore ensuring easy sterilization. This guarantees fault-free, precise and safe operation.