The Stomacher® Biomaster lab blenders offer fast reliable blending and homogenisation of a broad range of laboratory samples.
- Compact, space saving size
- Versatile preparation system for life science applications
- Maximise extraction
- Diluent-free sample processing
The Stomacher® machines work by the action of twin-reciprocating paddles on the sample contained inside a sterile plastic bag. The Stomacher® 80 Biomaster and Stomacher® 80 microBiomaster laboratory paddle blenders provide small tissue processing solutions for clinical and life science microbiology.
Stomacher® 80 technology provides fast, efficient and economical preparation of viable cell suspensions from tissue samples, with numerous methods documented for tissue biopsies, antibiotics, veterinary and forensic applications. Enhanced Stomacher® 80 bag range features closure and strainer bag formats, double bags for biosafety and the new microBag for effective homogenisation and sample recovery below 1 ml volumes.
Stomacher® Lab Blenders offer 5000 fold improvement in cell recovery and viability from traditional methods with dilutent-free sample processing. There is no cross contamination between samples, and processing time as well as paddle speed is adjustable.
Capable of processing samples from 250 µl to 80 ml, the Stomacher® 80 range caters for the enhanced sensitivity of downstream processing techniques that demand smaller samples without reducing sample quality.
Delivery information: Machine comes with three year warranty and ten year service support.