Stand Base Only, Cast Zinc-Aluminum Epoxy Coated, Caframo
Supplier: Caframo
A110BASE A110BASE
52428-764EA 344.56 CAD
52428-764 CA58960-156
Stand Base Only, Cast Zinc-Aluminum Epoxy Coated, Caframo
Stirrers Stirring Tools
Wide base and H configuration provide excellent stability for bench top stirrers, including high torque line - Models BDC1850 (CA58960-180), BDC3030 (CA58960-165), BDC6015 (CA58960-170), BDC2010 (CA97001-136) and BDC2002 (CA58960-174 ).
  • Wide base and H configuration – excellent stability
  • Cast zinc–aluminium alloy coated with chemical resistant epoxy paint (chemical and corrosion resistant – durable and easy to clean)
  • Rods interchangeable using same hex key – easy to adapt to process needs
This durable stand provides excellent stability for bench stop stirrers even when agitating viscous materials. The wide base and H configuration allow for a wide range of vessel sizes and use with hot plates. Portable and adaptable to address changing work spaces, equipment, and priorities. To be paired with ROD28, ROD38, ROD48, all interchangeable with this common base. The cast zinc-aluminum alloy base is epoxy coated for protection from corrosion and chemical spills.