VWR® Sharps Containers with Sliding Lid
Supplier: VWR International
76319-820 76319-822 76319-824
76319-820CS 494.54 CAD
76319-820 76319-822 76319-824
VWR® Sharps Containers with Sliding Lid
Bins Sharps Containers
Containers provide for the safe disposal of sharps and biohazard waste. The sliding lid design is perfect for busy professional settings such as laboratories, hospitals, dental, and physician offices requiring one-handed access to open sharps containers.
  • Torturous path containers are intended for use in a patient room or unsupervised area
  • Designed to prevent untrained hands from making contact with contents
  • Wall mountable and stackable
  • Designed for use in high activity environments
  • Translucent colors with lead-free pigments containing no heavy metals
  • Constructed of puncture and impact resistant material
  • Safe to autoclave or incinerate
Choose from one of three sharps containers with tortuous path lid designs. Lightweight and transportable, the containers are ideal for any professional that needs an easy yet safe way to dispose of biohazardous waste. A quick access rotating door on the lid lets users drop in waste and move the lid to a 'temporary close' position for storage between use. Safe to autoclave or incinerate.
Certifications: Meets or exceeds FDA standards including OSHA and EPA requirements for local biohazard waste collection.