Westcott® 180° Protractor Measuring Tools, Acme United
Supplier: Acme United
375
470121-020CS 667.2 CAD
470121-020
Westcott® 180° Protractor Measuring Tools, Acme United
Protractors
Westcott drafting and drawing tools are some of the finest products of their kind for students and professionals.
  • Semi-circular protractor: 4", 180°
  • Calibrated in 0.5"
  • Laminated for durability
  • Solid center
  • Translucent color allows for viewing through to the page
  • Perfect for a variety of drafting and drawing needs
A commitment to meticulous quality and cutting-edge innovation in measurement and art materials has earned an unparalleled reputation.