Neo-Clear® Xylene Substitute, HARLECO®, EMD Millipore
Supplier: EMD Millipore
Neo-Clear®
65351-85
CA34172-048CS 827.87 CAD
CA34172-048
Neo-Clear® Xylene Substitute, HARLECO®, EMD Millipore
Stains Microscopy and staining reagents
Solvent used as a medium for clearing and deparafinizing steps in staining and tissue processing
Aromatic solvent used as a medium for clearing and deparafinizing steps in staining and tissue processing. Use with Neo-Mount™ coverslipping medium, Catalog #65038-71.