VWR® Microcentrifuge Tubes, Low-Binding
Supplier: VWR
VWR®
1110-10-201AV 1310-10-301AV 1210-10-310AV
76332-068CS 886.39 CAD
76332-068 76332-058 76332-066
VWR® Microcentrifuge Tubes, Low-Binding
Tubes Microcentrifuge Tubes
These microcentrifuge tubes have a low adhesion surface which provides minimal to low binding. For use with DNA and viscous proteins, these non stick, graduated, polypropylene tubes may be used between –80 to +122 °C (–122 to +251.6 °F).
  • Leak proof snap caps
  • Autoclavable
  • Graduated markings every 100 µl
  • Rated to 20000×g
  • All tubes are nonsterile
  • Certified free of RNase, DNase and pyrogens
VWR® microcentrifuge tubes feature flat caps and frosted areas on the sides for easy labeling. Leakproof snap caps may be penetrated by a syringe. Easy-to-open caps will not pop open during autoclaving. The 0.5 ml tubes fit many thermal cyclers.