Constant Climate Chambers, KMF Series, BINDER
Supplier: Binder
9020-0342 9020-0344 9020-0346
76331-850EA 45526.59 CAD
76331-850 76331-862 76331-866
Constant Climate Chambers, KMF Series, BINDER
Testing Chambers Climatic Chambers
KMF constant climate chambers ensure constant test conditions throughout the stainless steel chamber even with a full load and only require a small amount of bench space. Units can be operated with pure aqua service, a flexible water purification system (optional) or deionized water. Units self-test for comprehensive status analysis.
  • APT.line™ preheating chamber technology
  • Humidity regulation with capacitive humidity sensor and vapor humidification
  • Internal data logger, measured values can be read out in open format via USB
  • Tight-sealing inner door made of safety glass (ESG)
  • Inner chamber made of stainless steel
  • Access port with silicone plug, 30 mm, left
  • Class 3.1 independent temperature safety device (DIN 12880) with visual and audible temperature alarm
The KMF series comes with an ethernet supported computer interface. The multi management software APT-COM™ basic edition makes the operation easy for the operator. Intuitive touchscreen controller with time-segment and real-time programming.
The humidification system requires minimal maintenance, it provides vapor pressure humidification for fast response times and has a drift-free, capacitive humidity sensor. All models have a powerful compressor cooling system for ambient temperatures up to 32 °C. The wide temperature and humidity range make this constant climate chamber ideal for stress testing series.
Values measured via the internal data logger can be read out in open format via USB. All models have an inner door made of ESG safety glass with a seal and double outer door seal, the door is heated against condensation. Glass corrosion is avoided thanks to the special TIMELESS coating. The KMF 240 and KMF720 units have four casters each, two with brakes on them.
Delivery information: Supplied with one stainless steel rack, test certificate and APT-COM™ three basic edition communication software. Units have an ethernet interface, a 30 mm Ø access port with silicone plug on the left and are supplied with safety connection kit for water supply and drainage, including 6 m water hose. A wide range of accessories are available and calibrations and validations are possible, for details please visit www.vwr.com or contact your local Avantor sales office.
Caution: Note suitable water quality:
Deionized water from an already existing on-site water treatment installation. Conductivity from 1 µS/cm up to a maximum of 20 µS/cm. (Ultrapure water which is in equilibrium with the CO2 in the air and has a conductivity of below 1 µS/cm may cause acid corrosion due to its low pH.)
Water treated by the optional pure aqua service, this system includes reusable measuring equipment to determine water quality.