Hematoxylin solution Gill III, HARLECO® used in nuclear staining, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: HydroxybrasilinDcH-48Natural Black I

65067-85 65067-75 65067-85 65067-75
CA15204-268EA 253.89 CAD
15204-268 CA15204-268 CA15204-182 15204-182
Hematoxylin solution Gill III, HARLECO® used in nuclear staining, Sigma-Aldrich®
Hematoxylin staining solutions

HARLECO® Triple strength nuclear stain used for routine tissue specimens glycol and methylmethacrylate sections. For regressive staining


  • Prepared from dyes certified by the Biological Stain Commission (BSC)
  • For In-Vitro Diagnostic Use
  • For Professional Use


Same as Gill I but triple the strength

Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078111
CAS Number: 517-28-2
IVDLogo

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR