Eosin Y (yellowish) alcoholic saturated solution, HARLECO® for histology and pathology, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Tetrabromofluorescein disodium saltEosin Y disodium saltEosin 2252′,4′,5′,7′-Tetrabromofluorescein disodium saltSolvent Red 43Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoateDisodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)Field's Stain B2,4,5,7-Tetrabromofluorescein disodium salt

586X-75 586X-75
CA15204-130EA 256.8 CAD
15204-130 CA15204-130
Eosin Y (yellowish) alcoholic saturated solution, HARLECO® for histology and pathology, Sigma-Aldrich®
Eosin Y (yellowish)

HARLECO® saturated solution of Eosin in alcohol. Used for glycol/methylmethacrylate processed tissues


  • Prepared from dyes certified by the Biological Stain Commission (BSC)
  • For In-Vitro Diagnostic Use.

Formula: C₂₀H₆Br₄Na₂O₅
MW: 691.86 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005040
CAS Number: 17372-87-1
IVDLogo

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR