PREempt® Concentrate Disinfectants, Contec
Supplier: Contec
11305 AHP1750CAN
CA10836-022CS 1435.07 CAD
CA10836-022 CA10789-317
PREempt® Concentrate Disinfectants, Contec
Cleaning Agents Disinfectants
One-step disinfectants cleaner utilizing Accelerated Hydrogen Peroxide®.
  • Concentrate solution is cost effective for daily use in large areas
  • Low residue and compatible with a wide range of materials
  • Odor-free for good operator acceptability
  • Does not require personal protection equipment (PPE) once diluted
  • Not classified as dangerous goods – no special shipping required
PREempt® concentrate is a patented synergistic blend of commonly used ingredients that when combined with low levels of hydrogen peroxide dramatically increase its potency and cleaningMore Product Information