PREempt™ CS 20 Disinfectant, Contec
Supplier: Contec
11401 AHP7CAN
CA10836-026CS 302.1 CAD
CA10836-026 CA10836-028 CA10836-036
PREempt™ CS 20 Disinfectant, Contec
Cleaning Agents Disinfectants
PREempt™ CS 20 is a sterilant and high level disinfectant for reprocessing heat sensitive semi-critical and critical devices for which heat sterilization is not suitable.
  • Chemical sterilant
  • Kills spores in 20 minutes
  • Contains hydrogen peroxide which breaks down into water and oxygen
  • Allows disinfection for non-autoclavable devices
  • 14 day reuse once opened
  • Only available in Canada
The exceptional safety profile of this 7% Accelerated Hydrogen Peroxide® (AHP®) solution means there is no special ventilation required. PREempt™ CS 20’s active Hydrogen Peroxide breaks down into water and oxygen, leaving no active ingredient residue.