Calcein AM 1 mg/mL in anhydrous DMSO fluorescent dye, for histology
Supplier: Biotium
Synonyms:
Calcein-AM
, Calcein O,O′-diacetate tetrakis(acetoxymethyl) ester
, Calcein acetoxymethyl ester
, Tetrakis(acetoxymethyl) 2,2',2'',2'''-(((3',6'-diacetoxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-2',7'-diyl)bis(methylene))bis(azanetriyl))tetraacetate
Calcein AM is a widely used green fluorescent cell marker. Calcein AM is membrane-permeant and thus can be introduced into cells via incubation.
- λEx/λEm (calcein) = 494/517 nm (pH 8)
- Colorless solid soluble in DMSO
- Store at -20°C and protect from light
Once inside the cells, calcein AM is hydrolyzed by endogenous esterase into the highly negatively charged green fluorescent calcein, which is retained in the cytoplasm. Calcein AM is an excellent tool for the studies of cell membrane integrity and for cell tracing. Supplied in lyophilized form or as a solution in anhydrous DMSO at 4 mM (80011-1) or 1 mg/mL (1 mM) (80011-2). Calcein AM is also offered in our Cell Viability and Cytotoxicity Assay Kit (30002).
Formula:
C₄₆H₄₆N₂O₂₃ MW: 994.86 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD05861516 CAS Number: 148504-34-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...