SATWipes® Delta™ Wipes, Contec®
Supplier: Contec
SAT-C3-694 SAT-C3-100 SAT-C3-7030 SAT-C3-7030/18 SAT-C3-8515
CA68300-358CS 507.5 CAD
CA68300-358 CA68310-578 CA68310-582 CA79999-888 10036-324
SATWipes® Delta™ Wipes, Contec®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
SATWipes® Delta™ wipes consist of a perforated roll of saturated wipes dispensed through a center-pull canister. The wipes are 100% nonwoven polyester fabric creating a soft, durable, and clean wipe.
  • Recommended for ISO Class 6 to 8 environments
  • Presaturated wipe eliminates the need for solvent bottles
  • Reusable dispenser reduces solvent handling
  • Different sizes and wipe quantities available for various customer needs
The isopropanol solvent blends provide safe cleaning solutions in various concentrations. Packaged with refill packs and a reusable pop-More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable