Hula Dancer basic Orbital Shaker, IKA
Supplier: IKA Works
0025003634 0025004320
76329-686EA 1296.12 CAD
76329-686 76329-818
Hula Dancer basic Orbital Shaker, IKA
Shakers Orbital Shakers
The Hula Dancers are powerful orbital shakers.
  • Robust housing
  • High stability
  • Touch function
Hula Dancer basic has a shaking diameter of 4.5 mm and allows a maximum load of 100 g. Speed is fixed at 2800 rpm.
Permissible relative humidity is 80%.
Certifications: IP protection class according to DIN EN 60529: IP 42.