PR Series Precision Balances, Ohaus
Supplier: Ohaus
30455673 30515026 30455605 30515027 30452494 30455606 30455607 30455670 30455671 30515038 30455668
76332-926EA 2585.78 CAD
76332-926 76332-976 76332-978 76332-980 76332-982 76332-984 76332-988 76332-992 76332-994 76332-996 76333-038
PR Series Precision Balances, Ohaus
Balances and Scales
The PR series of balances has been designed for routine weighing applications in the workplace, basic weighing, parts counting, percent weighing.
  • Equipped with three essential weighing modes
  • ABS housing, stainless steel pan, glass draftshield (milligram models only), weigh below hook, security lock interface, LFT lockout switch
  • All models have an easy-to-read, bright backlit liquid crystal display (LCD) and a simple user interface
  • PR is incredibly easy to operate, with almost no training required
Offering accuracy and repeatability in essential weighing applications in laboratory, industrial and education settings, PR Precision balances deliver competitive performance at an economical price.
Featuring RS-232 connectivity for data transfer and storage, and internal calibration (select models), making it ideal for routine weighing applications.
Offering user-selectable environmental filters and brightness settings, auto-tare, auto-dim, user-selectable span calibration points, software lockout and reset menu, userselectable communication settings and data print options, software overload/underload indicator, stability indicator.
Accessories information: Accessories not included.
Ordering information: Manufacturer’s two year parts and labor warranty (U.S. and Canada).