Roto-Mini™ Rotating Mixer, Benchmark Scientific
Supplier: BENCHMARK SCIENTIFIC CA
R2024 R2020
CA76324-002EA 1261.01 CAD
CA76324-002 CA76333-864
Roto-Mini™ Rotating Mixer, Benchmark Scientific
Shakers Rotating/Rocking Shakers
Roto-Mini™ series of rotating mixers provide a wide range of mixing capabilities, suitable for an array of laboratory applications including resuspension, protein binding, nucleic acid purification and more.
  • Interchangeable tube holders for accommodatng tube sizes from 0.5 ml to 50 ml
  • Adjustable motion from vertical to horizontal mixing
The Roto-Mini™ rotator is also available in a "Plus" configuration having additional features like variable speed from 5 to 70 rpm, providing mixed function like rotating in combination with oscillations for improved mixing, pause function to have intermittent stops during rotation, and rock function with back and forth movement and provides a gentle rocking motion.
The plus model also offers an additional rocking platform (4.5×3"), as well as a 96×0.2 ml tube/strip/plate holder for mixing PCR vessels in the mix (oscillating) mode.
Delivery information: Two years warranty.