BD Vacutainer® Blood Transfer Device, BD
Supplier: BD Medical
BD Vacutainer®
36488000
VT36488000CS 604.18 CAD
VT36488000
BD Vacutainer® Blood Transfer Device, BD
Blood Collection Tube Holders
The BD Vacutainer® blood transfer device (female) facilitates a safe transfer of blood from a syringe or blood culture bottle into a blood collection tube.
  • Sterile
  • Latex-free
  • Red colored tip for differentiation
The blood transfer device is designed with healthcare workers' safety in mind. It reduces the risk of transfer-related injuries while maintaining the specimen integrity required for accurate disease diagnosis.
Certifications: The device meets all procedural and safety standards for blood transfer from a conventional hypodermic needle and syringe to a blood collection tube or blood culture bottle.