6200 Series ESD Cleanroom Frocks, Polyester Fabric with Knit Cuffs, Transforming Technologies
Supplier: Transforming Technologies
JLM6201NB JLM6201WH JLM6202NB JLM6202WH JLM6203NB JLM6203WH JLM6206NB JLM6206WH JLM6207NB JLM6207WH JLM6208NB JLM6208WH JLM6209NB JLM6209WH JLM6204NB JLM6204WH JLM6205NB JLM6205WH
76329-320EA 148.2 CAD
76329-320 76329-322 76329-324 76329-326 76329-328 76329-330 76329-332 76329-334 76329-336 76329-338 76329-340 76329-342 76329-344 76329-346 76329-364 76329-366 76329-368 76329-370
6200 Series ESD Cleanroom Frocks, Polyester Fabric with Knit Cuffs, Transforming Technologies
Workwear
The ESD Cleanroom Frock is engineered to meet Class 100 cleanroom demands for industries such as microelectronics, semi-conductors, disk drives, laser, and similar industries. Cleanroom Frocks are made with static dissipative fabric to eliminate static on the wearer. Superior workmanship and reliable panel-to-panel continuity makes the 6200 ESD Cleanroom Frock an excellent product to control contamination and static.
  • Extremely comfortable and durable
  • Reliable panel-to-panel continuity tested after 100 washes
  • Knit cuff sleeve termination
  • Surface resistance: <106 Ω
Cleanroom frocks are designed to be worn in controlled environments where contamination is a concern. This frock is made to control static and does not generate a significant charge. It creates a Faraday cage effect and shields ESD susceptible items from charges on workers' clothing.
Superior workmanship and reliable panel-to-panel continuity makes the 6200 ESD Cleanroom Frock an excellent product to control contamination and static. The 6200 Series garments maintain consistent continuity readings for up to 100 wash cycles. The knee-length frocks protect the environment from the primary source of contamination found in cleanrooms - people. These frocks feature a military collar, ESD knit cuffs, and YKK zipper closure with adjustment snaps at the neck.
Certifications: Meets or exceeds requirements of ANSI ESD-S20.20.
Packaging: Each frock is packaged seperately.