pH 5 to 9 Test Strips, Precision Laboratories
Supplier: Precision Laboratories
PH5090-1B-50
470201-152EA 10.95 CAD
470201-152
pH 5 to 9 Test Strips, Precision Laboratories
Test Papers pH Indicator Papers and Test Strips
The pH 5 - 9 test strip is a narrow range strip and is ideal for weak solutions, such as alkaline or rain water, measuring in steps of 1.0 pH units.
  • Rapid test results
  • Distinct color changes for easier reading and more accurate results
To use, simply dip one strip into the test solution and remove. Shake off any excess liquid. Immediately compare to the color chart for results.
Packaging: Test strips are packaged in a clear, resealable 3×4" bag.