Polystyrene Spot Plates
Supplier: GSC INTERNATIONAL SE
1875-10
470313-468CS 214.05 CAD
470313-468
Polystyrene Spot Plates
Staining Tiles
Polystyrene spot plates have 12 depressions that are 20 mm in diameter by 10 mm in depth.
Overall plate dimension: 80×110 mm