Cover Glass with Fire-Polished Edge
Supplier: GSC INTERNATIONAL SE
000-109-10 000-110-10 000-105-10 000-112-10 000-111-10 000-107-10 000-108-10
470313-380CS 2053 CAD
470313-380 470313-378 470313-464 470313-376 470313-384 470313-374 470313-382
Cover Glass with Fire-Polished Edge
Watch Glasses
Made from borosilicate glass that resists cracking from temperature changes.
  • Borosilicate cover glasses
  • Fire-polished edges
  • Resistant to cracking from temperature changes
These borosilicate cover glasses have fire-polished edges and a concave shape. They are available in multiple diameters.