Eppendorf® Reference® 2 Multichannel Pipettors
Supplier: Eppendorf
4926000026 4926000034 4926000050 4926000042 4926000069 4926000018
10032-658EA 1967.9 CAD
10032-658 CA10032-520 CA10032-658 CA10032-660 CA10032-662 CA10032-724
Eppendorf® Reference® 2 Multichannel Pipettors
Pipettes Micropipettes
The unique one-button operation offers fast and ergonomic handling with reduced operating effort and active aerosol reduction.
The one-button operating system allows users to move more quickly through pipetting tasks with all operating controls (Aspiration/Dispensing, Volume Selection, Tip Ejection) contained in a single button. With the ability to eject a tip immediately following a dispensing step, aerosol contamination of the pipettor is considerably reduced. Releasing the operating button allows only fresh air, not residual aerosols, to re-enter the pipettor housing after the tip isMore Product Information