Bottles, Boston Round, Amber / Clear
Supplier: GSC International
405-5-DZ 407-2-GR 405-3-DZ 407-2-DZ 405-2-DZ 409-1-DZ 409-2-DZ 409-3-DZ 408-05-DZ 405-4-DZ 408-1-DZ 408-2-DZ 408-3-DZ 405-6-DZ 409-7-DZ 409-8-DZ 409-9-DZ 405-3D-DZ 405-3D-GR 405-7D-DZ
470205-806PK 138.25 CAD
470205-806 470146-176 470189-416 470206-124 470313-358 470313-362 470313-364 470313-366 470313-416 470313-456 470313-458 470313-460 470313-462 470313-494 470313-496 470313-498 470313-500 470325-202 470338-624 470338-666
Bottles, Boston Round, Amber / Clear
Bottles Dropping Bottles
The amber bottles with dropper assembly provide excellent UV protection for the content and the dropper allows for dispensing a small amount of liquid.
  • Amber colored glass provides UV protection
  • Provided with a dropper assembly or screw cap
  • Pipette is made of glass
  • Cap is made of plastic
  • Bulb is made of rubber
Amber flint glass boston round bottles are available in multiple sizes. Sold with a dropper assembly or a cap.
Packaging: Sold in packs of 12.