Porcelain Crucibles and Covers
Supplier: GSC International
CHF-30-LID-20 CLF-15-LID-20 CHF-10-20 CHF-15-20 CHF-20-20 CHF-50-LID-20 CLF-15-20 CLF-30-20 CLF-50-20 CHF-30-20 CHF-50-20 CHF-15-LID-20
470313-354PK 20.7 CAD
470313-354 470313-356 470313-396 470313-398 470313-400 470313-402 470313-404 470313-406 470313-408 470313-450 470313-452 470313-454
Porcelain Crucibles and Covers
Crucibles
These crucibles are great for student use.
  • High and low form available
Crucibles are uniform and dense. They have stable mechanical strength and little permeability, and they are available in both high form and low form. When properly used, the glazed, high form pieces withstand temperatures of 1050 °C. Unglazed, low form pieces will withstand temperatures of 1350 °C.
Ordering information: Replacement covers are also available.
Packaging: Sold in packs of 20.