Cryogenic Vials
Supplier: GSC International
CRYTB-1.8 CRYTB-5
470313-434CS 740.85 CAD
470313-434 470313-502
Cryogenic Vials
Vials
Cryogenic vials are used for storing biological samples at extreme temperatures.
  • Leak-proof
  • Printed graduations
  • White area for labeling
Our vials can withstand temperatures as low as –196 °C (approximately –321 °F) and have leak-proof silicone rubber lined caps, printed graduations, and a white area for labeling their contents.
1.8 ml vials measure 45.7 mm (1.8") tall and 13 mm (0.51") in diameter, and our 5 ml vials measure 56.9 mm (2.24") tall and 20 mm (0.79") in diameter.
Order Now
SPECIFICATIONS
Environmentally Preferable