Polypropylene Buchner Funnels
Supplier: GSC International
BFP100 BFP125 BFP70
470313-386EA 16.9 CAD
470313-386 470313-418 470313-476
Polypropylene Buchner Funnels
Funnels Filtration Funnels Büchner Funnels
The polypropylene Buchner funnels are two-piece, light-weight, and easy-to-clean.
They are fully autoclavable. The top portion snaps to the bottom forming a vacuum tight seal.
Order Now
SPECIFICATIONS
Environmentally Preferable