Porcelain Buchner Funnels
Supplier: GSC INTERNATIONAL SE
4-6054-10 4-6053-10 4-6052-10
470313-474EA 18.2 CAD
470313-474 470313-472 470313-470
Porcelain Buchner Funnels
Funnels Filtration Funnels Büchner Funnels
Cost-efficient, high quality Buchner Funnels.
  • Three sizes available
  • Fragile
These Buchner Funnels are made of a high-quality, laboratory-grade porcelain and have uniform wall thickness and dense materials to make a product with stable mechanical strength and little permeability. When properly used, the glazed pieces can withstand temperatures of 1050 °C and the unglazed pieces can withstand temperatures up to 1350 °C.