VWR® Syringe Pipette Tips, Evolution
Supplier: VWR International
ES40017-0007V ES40017-0011V ES40017-0012V ES40017-0013V ES40017-0014V ES40017-0015V ES40017-0016V ES40017-0017V ES40017-0000V ES40017-0001V ES40017-0003V ES40017-0004V ES40017-0005V ES40017-0006V
76321-574PK 313.93 CAD
76321-574 76321-576 76321-578 76321-580 76321-582 76321-584 76321-586 76321-588 76321-816 76321-818 76321-820 76321-822 76321-824 76321-826
VWR® Syringe Pipette Tips, Evolution
Pipette Tips
Evolution syringe tips are compatible with most repeating pipettors on the market. Positive displacement provides precise dosing at even the smallest volumes.
  • Syringe tips come with a color coded adapter for quick identification of the correct size tip
  • Seven tip sizes dispense volumes from 1 µl to 10 ml
  • Available sterile and non sterile
All sizes of the Evolution syringe tips provide reliable and repeatable dosing for a wide range of liquids including viscous, volatile, and infectious material. Sterile tips are individually wrapped and certified to be pyrogen, ATP, RNase, and DNase free. Color coded adapters allow for quick identification of the correct size tip. Manufactured from virgin grade polypropylene.