HPTLC Plates, Nano Silica Gel 60, Unmodified Layers, Nano-SIL, Macherey Nagel
Supplier: Macherey-Nagel
811012 811022
29444-518PK 1083.34 CAD
29444-518 29444-520
HPTLC Plates, Nano Silica Gel 60, Unmodified Layers, Nano-SIL, Macherey Nagel
TLC Plates
Unmodified HPTLC silica layers for HPTLC with nano silica gel (particle size 2 to 10 μm).
  • Specific surface (BET) ~500 m2/g
  • Specific pore volume 0.75 ml/g
  • Narrow fractionation of silica particles
Binders are stable in almost all organic solvents. The indicatior is manganese that activate zinc silicate with green fluorescence for short-wave UV (254 nm).
Allows sharper separations, shorter developing times and an increased detection sensitivity compared to standard silica gel plates.
Order Now
SPECIFICATIONS
Environmentally Preferable