VWR® 96-Well Deep-Well Plates with Automation Notches
Supplier: VWR
VWR®
775A-S0-621AV 775B-S0-621AV 775A-00-621AV 775B-00-621AV
76330-000CS 762.45 CAD
76330-000 76330-002 76329-998 76329-996
VWR® 96-Well Deep-Well Plates with Automation Notches
Microplates Deep Well Microplates
Heavy polypropylene deep well plates feature square wells, molded alphanumeric labeling, and smooth interior surfaces to ensure that all the samples can be pipetted.
  • Alphanumeric indexing
  • Square well with V-bottom design
  • Autoclavable
  • Freezable to –80 °C (–112 °F)
VWR® deep-well plates feature printed alphanumeric labeling and smooth interior surfaces to ensure easy pipetting. Plates and mats are free of RNase, DNase and human DNA (lot-by-lot certificates available).
Applications include HTS, combinatorial chemistry, library applications (compound storage), immunology (serial dilutions), cell culture (yeast growth, bacteria, T-Cells), DMPK (use with filtered plates on manifold), mini prep work, and master mixing for Next Gen sequencing.
They are designed to be manipulated by robotic systems and can be sealed using film or mats. The front notches are compatible with some Hamilton® platforms.